Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456081_m1
          See other H2-T24 GE Assays ›
          Gene Symbol:
          H2-T24
          Gene Name:
          histocompatibility 2, T region locus 24
          Gene Aliases:
          H-2T24
          Chromosome Location:
          Chr.17: 36003992 - 36020842 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456081_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-T24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008207.3 NP_032233.2 3 - 4 674 89
          GenBank mRNA L22338.1 - 3 - 4 628 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001993.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-T24

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003996 NM_008207.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003991 NM_008207.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003991 L22338.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141218 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003996 NM_008207.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003991 NM_008207.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003991 L22338.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P581082 NM_008207.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P581082 L22338.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189812 NM_008207.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P189812 L22338.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31162242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03312638 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08769830 - -
          Whole Mouse Genome G4122A A_52_P581082 NM_008207.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581082 L22338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189812 NM_008207.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P189812 L22338.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08769830 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03312638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13409450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13503238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13258988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13383251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13450451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13578144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13258990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13549584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13377015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13578143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13578147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13549590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13534120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13167227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13338385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13549586 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450676 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450678 - -
          Affymetrix HT MG-430 PM Array Plate 1422160_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450675 - -
          Mouse Genome 430 2.0 Array 1422160_PM_at - -
          Mouse Genome 430 2.0 Array 1422160_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15042_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15042_at - -
          Affymetrix Mouse Clariom S Array PSR1700019067.mm.1 NM_008207.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019067.mm.1 NM_008207.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019067.mm.1 TC1700001993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019068.mm.1 NM_008207.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019068.mm.1 NM_008207.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019068.mm.1 TC1700001993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019069.mm.1 NM_008207.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019069.mm.1 NM_008207.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019069.mm.1 TC1700001993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019065.mm.1 NM_008207.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019065.mm.1 NM_008207.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019065.mm.1 TC1700001993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019066.mm.1 NM_008207.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019066.mm.1 NM_008207.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019066.mm.1 TC1700001993.mm.1 MTA TC
          Mouse Expression Set 430 1422160_at - -
          Mouse Expression Set 430 1422160_PM_at - -
          Murine Genome U74v2 Set 94746_at - -
          Murine Genome U74v2 Set 94746_PM_at - -
          Mouse Genome 430A 2.0 Array 1422160_at - -
          Mouse Genome 430A 2.0 Array 1422160_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422160_at - -
          Affymetrix HT Mouse Genome 430A Array 1422160_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6370402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613823 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613823 - -
          mouse-6 v1.1 (Illumina) scl0015042.1_73-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015042.1_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229173 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229173 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline