Search
Search
| Gene Symbol: Orc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159563.1 | NP_001153035.1 | 5 - 6 | 482 | 93 | |
| NM_015824.4 | NP_056639.3 | 5 - 6 | 482 | 93 | ||
| GenBank mRNA | AB190257.2 | - | 5 - 6 | 449 | 93 | |
| AJ132830.1 | - | 5 - 6 | 464 | 93 | ||
| AK045388.1 | - | 5 - 6 | 457 | 93 | ||
| AK086579.1 | - | 5 - 6 | 485 | 93 | ||
| AK145324.1 | - | 5 - 6 | 461 | 93 | ||
| BB663298.1 | - | 5 - 6 | 482 | 93 | ||
| BC061252.1 | - | 5 - 6 | 453 | 30250245 | 93 | |
Set Membership: |
|