Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457483_s1
          See other MC4R GE Assays ›
          Gene Symbol:
          Mc4r
          Gene Name:
          melanocortin 4 receptor
          Gene Aliases:
          Mc4-r
          Chromosome Location:
          Chr.18: 66857705 - 66860487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457483_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mc4r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016977.4 NP_058673.2 1 501 79
          GenBank mRNA AK136793.1 - 1 487 79
          BC116957.1 - 1 119 40126276 79
          BC116959.1 - 1 119 40126278 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001485.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mc4r

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          diet induced thermogenesis
          energy reserve metabolic process
          signal transduction
          G-protein coupled receptor signaling pathway
          feeding behavior
          regulation of metabolic process
          insulin secretion
          positive regulation of cAMP biosynthetic process
          response to insulin
          positive regulation of bone resorption
          regulation of feeding behavior
          negative regulation of feeding behavior
          regulation of grooming behavior
          signal transducer activity
          G-protein coupled receptor activity
          melanocortin receptor activity
          melanocyte-stimulating hormone receptor activity
          protein binding
          peptide hormone binding
          ubiquitin protein ligase binding
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 AK136793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 BC116957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2183884 NM_016977.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185714 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147350 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 AK136793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 BC116957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2183884 NM_016977.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 AK136793.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 BC116957.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 BC116959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268709 NM_016977.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31788800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147350 - -
          Whole Mouse Genome G4122A A_51_P268709 AK136793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 BC116957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 BC116959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268709 NM_016977.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117217 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122164 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147350 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14102913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31788759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14000913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13976333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14102955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13963845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13963830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14000912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31788783 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10459517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459515 - -
          Affymetrix Mouse Gene 1.1 ST Array 10459512 - -
          Affymetrix Mouse Clariom S Array PSR1800010952.mm.1 NM_016977.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010952.mm.1 TC1800001485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010951.mm.1 NM_016977.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010951.mm.1 TC1800001485.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 366456 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 430333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2611237 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2611237 - -
          mouse-6 v1.1 (Illumina) scl14872.1.1_153-S - -
          Illumina Sentrix mouse-6.v1 bead array 430333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14872.1.1_153-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2611237 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline