| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00003545.mm.1 |
NM_028242.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003545.mm.1 |
NM_028242.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003545.mm.1 |
TC0X00000530.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00003548.mm.1 |
NM_028242.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003548.mm.1 |
NM_028242.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003548.mm.1 |
NM_029371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003548.mm.1 |
NM_029371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003548.mm.1 |
TC0X00000530.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00003544.mm.1 |
NM_028242.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003544.mm.1 |
NM_028242.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00003544.mm.1 |
TC0X00000530.mm.1 |
MTA TC |