| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569629 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
200587 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
813651 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
832538 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861004 |
AK078838.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861004 |
BC032265.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861004 |
NM_181420.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946048 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
772025 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
271394 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
330264 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
152762 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649441 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93518 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327497 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
744040 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
36432 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
26142 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
64681 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149963 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166256 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452941 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452948 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452945 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452940 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452953 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452960 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452949 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452956 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452959 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452962 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452963 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452964 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452935 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452938 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452955 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30452951 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149963 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08467079 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03105075 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08467094 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166256 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08467152 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK139992.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
BC051676.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK028837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
AK029028.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P166266 |
NM_029926.5 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
BC051676.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK028837.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK029028.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
AK078803.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P67440 |
NM_029926.5 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108536 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110430 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149963 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166256 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12412293 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12054856 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12309945 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12412303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453105 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452973 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453019 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452981 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452963 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453106 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453053 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452978 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453060 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452960 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453107 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453098 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30452998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30453050 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452935 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452948 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452938 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452962 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12330726 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452965 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452941 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12330727 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452956 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452955 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452963 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12077553 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452960 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452944 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452966 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452964 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452934 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452954 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452957 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452953 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30452940 |
- |
- |