Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459541_m1
          See other TEX13 GE Assays ›
          Gene Symbol:
          Tex13
          Gene Name:
          testis expressed gene 13
          Gene Aliases:
          4933403J23Rik, Tex13b
          Chromosome Location:
          Chr.X: 140808307 - 140813451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459541_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031381.2 NP_113558.1 2 - 3 542 70
          GenBank mRNA AF285576.1 - 2 - 3 542 70
          BC116863.1 - 2 - 3 518 40126182 70
          BC119125.1 - 2 - 3 518 8733874 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003079.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex13

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P648433 NM_031381.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P648433 AF285576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P648433 NM_031381.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P648433 AF285576.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58710 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178757 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P648433 NM_031381.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P648433 AF285576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P648433 NM_031381.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P648433 AF285576.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32701065 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178757 - -
          Whole Mouse Genome G4122A A_52_P648433 NM_031381.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P648433 AF285576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P648433 NM_031381.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P648433 AF285576.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100902 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14994752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14994747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14994756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14912587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14994741 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14994746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32700995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32700991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32701028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15164709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15196933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32700993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15098682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15142049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14708685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14845615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15142050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32700989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14912587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32700986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32700991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15196929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15142047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15196937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15142037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32701026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606998 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607000 - -
          Affymetrix HT MG-430 PM Array Plate 1450555_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432159_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606997 - -
          Mouse Genome 430 2.0 Array 1432159_a_at - -
          Mouse Genome 430 2.0 Array 1432159_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450555_PM_at - -
          Mouse Genome 430 2.0 Array 1450555_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83555_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83555_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00019341.mm.1 NM_031381.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019341.mm.1 NM_031381.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019341.mm.1 TC0X00003079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019343.mm.1 NM_031381.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019343.mm.1 NM_031381.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019343.mm.1 TC0X00003079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019344.mm.1 NM_031381.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019344.mm.1 NM_031381.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019344.mm.1 TC0X00003079.mm.1 MTA TC
          Mouse Expression Set 430 1450555_at - -
          Mouse Expression Set 430 1450555_PM_at - -
          Mouse Expression Set 430 1432159_PM_a_at - -
          Mouse Expression Set 430 1432159_a_at - -
          Mouse Genome 430A 2.0 Array 1432159_a_at - -
          Mouse Genome 430A 2.0 Array 1450555_at - -
          Mouse Genome 430A 2.0 Array 1450555_PM_at - -
          Mouse Genome 430A 2.0 Array 1432159_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432159_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432159_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450555_at - -
          Affymetrix HT Mouse Genome 430A Array 1450555_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 326590 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780093 - -
          Illumina mouse-6 v1.0 expression beadchip 780093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258183 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258183 - -
          mouse-6 v1.1 (Illumina) scl53787.4.1_142-S - -
          Illumina Sentrix mouse-6.v1 bead array 780093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53787.4.1_142-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2850784 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2850784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline