Search
Search

Rbfox2| Gene Symbol: Rbfox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110827.2 | NP_001104297.1 | 10 - 11 | 1604 | 78 | |
| NM_001110828.2 | NP_001104298.1 | 9 - 10 | 1158 | 78 | ||
| NM_001110830.2 | NP_001104300.1 | 9 - 10 | 1146 | 78 | ||
| NM_001286418.1 | NP_001273347.1 | 6 - 7 | 864 | 78 | ||
| NM_053104.6 | NP_444334.3 | 10 - 11 | 1616 | 78 | ||
| GenBank mRNA | AF229055.1 | - | 9 - 10 | 1089 | 78 | |
| AF387322.1 | - | 9 - 10 | 890 | 78 | ||
| AK077594.1 | - | 10 - 11 | 1492 | 78 | ||
| AY659951.1 | - | 9 - 10 | 890 | 78 | ||
| AY659953.1 | - | 6 - 7 | 608 | 78 | ||
| AY904025.1 | - | 11 - 12 | 1040 | 78 | ||
| BC002124.1 | - | 9 - 10 | 984 | 3494630 | 78 | |
| DQ017388.1 | - | 9 - 10 | 902 | 78 | ||
Set Membership: |
|