Search
Search

Zmynd10| Gene Symbol: Zmynd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053253.3 | NP_444483.2 | 10 - 11 | 1267 | 65 | |
| GenBank mRNA | AF123386.1 | - | 8 - 9 | 898 | 65 | |
| AK132332.1 | - | 10 - 11 | 1276 | 65 | ||
| BC052357.1 | - | 10 - 11 | 1267 | 6772144 | 65 | |
Set Membership: |
|