| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574695 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
99358 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
207984 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
694521 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379148 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449954 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
663986 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910993 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
544122 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478104 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
61838 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347011 |
BC117048.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347011 |
BC117050.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347011 |
NM_011463.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347011 |
Y11505.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446077 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516595 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
180113 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
663804 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
430658 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
127676 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
169743 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P141787 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P170959 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177127 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177126 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177143 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177138 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177137 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177147 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177153 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177128 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177121 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177134 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177130 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177146 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177140 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177136 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177151 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177123 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177149 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28177131 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07513160 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02444307 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02444287 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02444267 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07513196 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P170959 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P141787 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
BC004692.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
BC029102.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
AK028888.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
AF363457.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P137611 |
NM_130886.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109694 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109189 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P141787 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P170959 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177315 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177271 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177094 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177326 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177290 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177111 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177344 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177149 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177305 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177172 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177248 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177146 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177321 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177309 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177107 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177154 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177262 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177350 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177311 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177096 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177272 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177343 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177332 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177289 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177360 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177263 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177297 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177287 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177347 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177260 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177286 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177314 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177274 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177228 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177251 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177213 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177301 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177217 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177300 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177324 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177377 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177277 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177292 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177298 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177302 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177369 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177357 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177143 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177319 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177175 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177233 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177299 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177353 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177304 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177313 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177308 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177340 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177363 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177258 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177280 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177367 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28177358 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177146 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177131 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177119 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09218994 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177158 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177149 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177134 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177147 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177144 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177123 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177136 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09518498 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177126 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177137 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09048165 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177127 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177138 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177111 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177151 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09245976 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177128 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177162 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177154 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177135 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08994293 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177120 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177116 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177143 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09121151 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09142957 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177140 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177166 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177109 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28177113 |
- |
- |