Search
Search
| Gene Symbol: Bco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133217.3 | NP_573480.1 | 4 - 5 | 647 | 70 | |
| GenBank mRNA | AJ290392.1 | - | 3 - 4 | 490 | 70 | |
| BC107006.2 | - | 3 - 4 | 490 | 40046913 | 70 | |
| BC107007.2 | - | 3 - 4 | 490 | 40046914 | 70 | |
Set Membership: |
|