Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460119_m1
          See other INSIG2 GE Assays ›
          Gene Symbol:
          Insig2
          Gene Name:
          insulin induced gene 2
          Gene Aliases:
          2900053I11Rik, C730043J18Rik, Insig-2
          Chromosome Location:
          Chr.1: 121304353 - 121333654 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460119_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insig2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133748.2 NP_598509.1 1 - 2 331 86
          GenBank mRNA AF527631.1 - 1 - 2 65 86
          AK152076.1 - 1 - 2 61 86
          AK154543.1 - 1 - 2 94 86
          AK161256.1 - 1 - 2 99 86
          AK165634.1 - 1 - 2 100 86
          AY156085.1 - 1 - 2 65 86
          BC023067.1 - 1 - 2 69 5357599 86
          BC024411.1 - 1 - 2 83 4971297 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002969.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF527631.1,AK152076.1,AK154543.1,AK161256.1,AK165634.1,BC023067.1,BC024411.1,NM_133748.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insig2

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          triglyceride metabolic process
          cholesterol biosynthetic process
          response to sterol depletion
          steroid metabolic process
          cholesterol metabolic process
          negative regulation of steroid biosynthetic process
          sterol biosynthetic process
          SREBP signaling pathway
          inner ear morphogenesis
          middle ear morphogenesis
          negative regulation of fatty acid biosynthetic process
          palate development
          cranial suture morphogenesis
          protein binding
          transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100023939.mm.1 NM_001271532.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023939.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023939.mm.1 TC0100002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023925.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023925.mm.1 NM_178082.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023925.mm.1 TC0100002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023924.mm.1 NM_001271531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023924.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023924.mm.1 NM_178082.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023924.mm.1 TC0100002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023932.mm.1 NM_001271532.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023932.mm.1 NM_133748.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023932.mm.1 TC0100002969.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 603014 - -

          Back To Top
           

          Citations (1)

          Fibroblast growth factor 21 corrects obesity in mice.
          Authors
          Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
          Journal
          Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 UTC 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline