Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460290_m1
          See other SLC39A3 GE Assays ›
          Gene Symbol:
          Slc39a3
          Gene Name:
          solute carrier family 39 (zinc transporter), member 3
          Gene Aliases:
          AI845814, Zip3
          Chromosome Location:
          Chr.10: 81028540 - 81037421 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460290_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134135.1 NP_598896.1 1 - 2 301 91
          GenBank mRNA AK087519.1 - 3 - 4 458 91
          BC005502.1 - 1 - 2 301 3490154 91
          BC085175.1 - 3 - 4 441 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002523.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a3

          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          in utero embryonic development
          transport
          ion transport
          zinc II ion transport
          metal ion transport
          T cell homeostasis
          embryonic cranial skeleton morphogenesis
          transmembrane transport
          limb development
          zinc II ion transmembrane transport
          zinc ion transmembrane transporter activity
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286996 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286999 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286998 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286997 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287002 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020143 BC085175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020143 BC005502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020143 NM_134135.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527219 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126428 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020143 BC085175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020143 BC005502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020143 NM_134135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P387248 BC085175.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P387248 BC005502.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P387248 AK087519.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P387248 NM_134135.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27287004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286980 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126428 - -
          Whole Mouse Genome G4122A A_51_P387248 BC085175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387248 BC005502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387248 AK087519.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387248 NM_134135.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103005 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08595580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08595579 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08626922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08869442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286999 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287002 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286997 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286996 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286998 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27287001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27287052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08903428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08460970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08460982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08460950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08626923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08460977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08793231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08595579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08746887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08732484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08869442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08746896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08460962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08825287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08327657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08930405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27287040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371038 - -
          Affymetrix HT MG-430 PM Array Plate 1417882_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417881_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371037 - -
          Mouse Genome 430 2.0 Array 1417881_PM_at - -
          Mouse Genome 430 2.0 Array 1417882_PM_at - -
          Mouse Genome 430 2.0 Array 1417882_at - -
          Mouse Genome 430 2.0 Array 1417881_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106947_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106947_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000018836.mm.1 NM_134135.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018836.mm.1 NM_134135.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018836.mm.1 TC1000002523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018837.mm.1 NM_134135.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018837.mm.1 NM_134135.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018837.mm.1 TC1000002523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018835.mm.1 NM_134135.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018835.mm.1 NM_134135.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018835.mm.1 TC1000002523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018838.mm.1 NM_134135.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018838.mm.1 NM_134135.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018838.mm.1 TC1000002523.mm.1 MTA TC
          Mouse Expression Set 430 1417882_at - -
          Mouse Expression Set 430 1417881_PM_at - -
          Mouse Expression Set 430 1417882_PM_at - -
          Mouse Expression Set 430 1417881_at - -
          Murine Genome U74v2 Set 93173_PM_at - -
          Murine Genome U74v2 Set 93173_at - -
          Murine Genome U74v2 Set 165796_at - -
          Murine Genome U74v2 Set 165796_PM_at - -
          Mouse Genome 430A 2.0 Array 1417881_at - -
          Mouse Genome 430A 2.0 Array 1417882_at - -
          Mouse Genome 430A 2.0 Array 1417882_PM_at - -
          Mouse Genome 430A 2.0 Array 1417881_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417881_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417882_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417881_at - -
          Affymetrix HT Mouse Genome 430A Array 1417882_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 724987 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3990397 - -
          MouseWG-6 v2 Expression BeadChip 4670494 - -
          Illumina mouse-6 v1.0 expression beadchip 4670494 - -
          Illumina mouse-6 v1.0 expression beadchip 3990397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695367 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2664375 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2664375 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2695367 - -
          mouse-6 v1.1 (Illumina) scl0003912.1_123-S - -
          mouse-6 v1.1 (Illumina) scl0106947.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990397 - -
          Illumina Sentrix mouse-6.v1 bead array 4670494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0106947.1_49-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003912.1_123-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670494 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2977240 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2695367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2977240 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0106947.1_49-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline