Search
Search

Kdm4c| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172095.1 | NP_001165566.1 | 6 - 7 | 844 | 61 | |
| NM_144787.2 | NP_659036.1 | 6 - 7 | 908 | 61 | ||
| GenBank mRNA | AK042792.1 | - | 5 - 6 | 623 | 61 | |
| AK048092.1 | - | 6 - 7 | 821 | 61 | ||
| AK053104.1 | - | 6 - 7 | 784 | 61 | ||
| AK081769.1 | - | 6 - 7 | 851 | 61 | ||
| AK144103.1 | - | 6 - 7 | 871 | 61 | ||
| AK144890.1 | - | 6 - 7 | 882 | 61 | ||
| AK173023.1 | - | 6 - 7 | 908 | 61 | ||
| BC020180.1 | - | 6 - 7 | 850 | 4923943 | 61 | |
| BC042424.1 | - | 6 - 7 | 844 | 5357211 | 61 | |
Set Membership: |
|