Search
Search

Camk1g| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144817.2 | NP_659066.1 | 4 - 5 | 434 | 71 | |
| GenBank mRNA | AF428262.1 | - | 4 - 5 | 382 | 71 | |
| AY212936.1 | - | 3 - 4 | 314 | 71 | ||
| BC021840.1 | - | 4 - 5 | 383 | 4502479 | 71 | |
| BY730299.1 | - | 4 - 5 | 435 | 71 | ||
Set Membership: |
|