Search
Search
| Gene Symbol: Oas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145226.2 | NP_660261.1 | 11 - 12 | 2588 | 70 | |
| GenBank mRNA | AB067534.1 | - | 11 - 12 | 2588 | 70 | |
| AF453830.1 | - | 11 - 12 | 2580 | 70 | ||
| BC016461.1 | - | 4 - 5 | 686 | 70 | ||
| BC141055.1 | - | 11 - 12 | 2581 | 9055667 | 70 | |
Set Membership: |
|