Search
Search
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145475.4 | NP_663450.3 | 9 - 10 | 1543 | 74 | |
| GenBank mRNA | AB079067.1 | - | 9 - 10 | 1050 | 74 | |
| AK042077.1 | - | 9 - 10 | 1134 | 74 | ||
| AK052269.1 | - | 9 - 10 | 1309 | 74 | ||
| AK129416.1 | - | 6 - 7 | 685 | 74 | ||
| AK161801.1 | - | 381 | 74 | |||
| AK169528.1 | - | 9 - 10 | 1128 | 74 | ||
| BC094253.1 | - | 9 - 10 | 1542 | 6400872 | 74 | |
Set Membership: |
|