Search
Search

Tinf2| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145705.3 | NP_663751.2 | 3 - 4 | 606 | 64 | |
| GenBank mRNA | AF214013.1 | - | 3 - 4 | 597 | 64 | |
| AF518764.1 | - | 3 - 4 | 571 | 64 | ||
| AK016456.1 | - | 3 - 4 | 539 | 64 | ||
| AK028952.1 | - | 3 - 4 | 577 | 64 | ||
| AK090324.1 | - | 3 - 4 | 573 | 64 | ||
| AK144592.1 | - | 3 - 4 | 535 | 64 | ||
| AK169480.1 | - | 1 - 2 | 121 | 64 | ||
| BC017142.1 | - | 1 - 2 | 794 | 64 | ||
| BC030347.1 | - | 3 - 4 | 532 | 5003804 | 64 | |
Set Membership: |
|