Search
Search

Abca8a| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 16 - 17 | 2212 | 89 | |
| GenBank mRNA | AB255881.1 | - | 15 - 16 | 2131 | 89 | |
| AB255882.1 | - | 16 - 17 | 2219 | 89 | ||
| AF498360.1 | - | 15 - 16 | 2140 | 89 | ||
| AY732492.1 | - | 15 - 16 | 2140 | 89 | ||
Set Membership: |
|