Search
Search

Tfap2d| Gene Symbol: Tfap2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153154.2 | NP_694794.1 | 3 - 4 | 833 | 93 | |
| GenBank mRNA | AF421891.1 | - | 3 - 4 | 860 | 93 | |
| AK132112.1 | - | 3 - 4 | 835 | 93 | ||
| BC152324.1 | - | 3 - 4 | 782 | 40141049 | 93 | |
| BC152325.1 | - | 3 - 4 | 782 | 40141051 | 93 | |
Set Membership: |
|