Search
Search

Wdr19| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153391.2 | NP_700440.2 | 30 - 31 | 3414 | 90 | |
| GenBank mRNA | AK147970.1 | - | 30 - 31 | 3414 | 90 | |
| AY029258.1 | - | 30 - 31 | 3484 | 90 | ||
| BC046432.1 | - | 15 - 16 | 1616 | 90 | ||
| BC082301.1 | - | 28 - 29 | 4846 | 90 | ||
Set Membership: |
|