Search
Search

BC028528| Gene Symbol: BC028528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153513.2 | NP_705733.2 | 2 - 3 | 133 | 107 | |
| GenBank mRNA | AF534879.1 | - | 2 - 3 | 133 | 107 | |
| BC028528.1 | - | 2 - 3 | 151 | 1366537 | 107 | |
Set Membership: |
|