Search
Search

Thoc1| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153552.3 | NP_705780.1 | 11 - 12 | 930 | 96 | |
| GenBank mRNA | AK031785.1 | - | 11 - 12 | 931 | 96 | |
| AK032200.1 | - | 11 - 12 | 931 | 96 | ||
| AK042867.1 | - | 11 - 12 | 936 | 96 | ||
| AK047991.1 | - | 7 - 8 | 2193 | 96 | ||
| AK052835.1 | - | 11 - 12 | 926 | 96 | ||
| AK139307.1 | - | 11 - 12 | 936 | 96 | ||
| AK167374.1 | - | 11 - 12 | 944 | 96 | ||
| AK168619.1 | - | 11 - 12 | 1195 | 96 | ||
| BC024951.1 | - | 11 - 12 | 4010605 | 96 | ||
| BC027149.1 | - | 5 - 6 | 512 | 96 | ||
Set Membership: |
|