Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463704_m1
          See other RRNAD1 GE Assays ›
          Gene Symbol:
          Rrnad1
          Gene Name:
          ribosomal RNA adenine dimethylase domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 87922601 - 87930588 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463704_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rrnad1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153562.4 NP_705790.3 5 - 6 866 89
          GenBank mRNA AK030684.1 - 4 - 5 986 89
          AK035389.1 - 5 - 6 818 89
          AK082223.1 - 5 - 6 799 89
          BC012224.1 - 5 - 6 758 89
          BC023814.1 - 4 - 5 706 5353932 89
          BC094052.1 - 3 - 4 1570 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002294.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rrnad1

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310775 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310773 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC094052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC012224.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC023814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 AK172049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 AK082223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC094052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC012224.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 BC023814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 AK172049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495162 AK082223.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83544 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149387 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC094052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC012224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC023814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 AK172049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 AK082223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC094052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC012224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 BC023814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 AK172049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495162 AK082223.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22310755 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04979467 - -
          Whole Mouse Genome G4122A A_52_P495162 BC094052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 BC012224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 BC023814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 AK172049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 AK082223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 BC094052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 BC012224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 BC023814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 AK172049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495162 AK082223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221739 BC012224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221739 AK082223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221739 AK035389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221739 NM_153562.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221739 AK167623.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03124993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02733437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02733439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310775 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310778 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22310773 - -
          Agilent Mouse cDNA Microarray (G4104A) 619118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22310745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02733434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02733437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02440641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02440637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02440642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02440638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03124993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22310740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499281 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493102 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499280 - -
          Affymetrix HT MG-430 PM Array Plate 1433961_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499273 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493094 - -
          Mouse Genome 430 2.0 Array 1433961_PM_at - -
          Mouse Genome 430 2.0 Array 1433961_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229503_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229503_at - -
          Affymetrix Mouse Clariom S Array PSR0300017309.mm.1 NM_153562.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017309.mm.1 NM_153562.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017309.mm.1 TC0300002294.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017306.mm.1 NM_153562.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017306.mm.1 NM_153562.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017306.mm.1 TC0300002294.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017307.mm.1 NM_153562.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017307.mm.1 NM_153562.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017307.mm.1 TC0300002294.mm.1 MTA TC
          Mouse Expression Set 430 1433961_at - -
          Mouse Expression Set 430 1433961_PM_at - -
          Murine Genome U74v2 Set 166299_s_at - -
          Murine Genome U74v2 Set 166299_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1433961_PM_at - -
          Mouse Genome 430A 2.0 Array 1433961_at - -
          Affymetrix HT Mouse Genome 430A Array 1433961_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433961_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 929001 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2120717 - -
          Illumina mouse-6 v1.0 expression beadchip 104070390 - -
          Illumina mouse-6 v1.0 expression beadchip 2120717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228168 - -
          mouse-6 v1.1 (Illumina) scl21989.6_25-S - -
          mouse-6 v1.1 (Illumina) scl00229503.2_106-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120717 - -
          Illumina Sentrix mouse-6.v1 bead array 104070390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00229503.2_106-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104070390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220593 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline