Search
Search

Prkdc| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 36 - 37 | 4793 | 74 | |
| GenBank mRNA | AB007544.1 | - | 36 - 37 | 4770 | 74 | |
| AB011543.1 | - | 36 - 37 | 4782 | 74 | ||
| D87521.1 | - | 36 - 37 | 4770 | 74 | ||
Set Membership: |
|