Search
Search
| Gene Symbol: Rrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145620.4 | NP_663595.1 | 11 - 12 | 67 | ||
| GenBank mRNA | AF368232.1 | - | 11 - 12 | 67 | ||
| AK076113.1 | - | 11 - 12 | 67 | |||
| AK156199.1 | - | 10 - 11 | 1150 | 67 | ||
| BC014703.1 | - | 11 - 12 | 1076 | 4237895 | 67 | |
Set Membership: |
|