Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00467539_m1
          See other NOD2 GE Assays ›
          Gene Symbol:
          Nod2
          Gene Name:
          nucleotide-binding oligomerization domain containing 2
          Gene Aliases:
          ACUG, BLAU, CD, Card15, F830032C23Rik, IBD1, Nlrc2
          Chromosome Location:
          Chr.8: 88647280 - 88688474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00467539_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nod2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145857.2 NP_665856.2 7 - 8 2703 69
          GenBank mRNA AF520774.1 - 7 - 8 2748 69
          AK089843.1 - 7 - 8 2667 69
          AY160220.1 - 6 - 7 2632 69
          AY160221.1 - 7 - 8 2704 69
          BC044774.2 - 7 - 8 2730 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001037.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nod2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          activation of immune response
          cytokine production involved in immune response
          cytokine secretion involved in immune response
          immune system process
          immunoglobulin production involved in immunoglobulin mediated immune response
          positive regulation of dendritic cell antigen processing and presentation
          negative regulation of T cell mediated immunity
          positive regulation of dendritic cell cytokine production
          positive regulation of type 2 immune response
          negative regulation of inflammatory response to antigenic stimulus
          positive regulation of humoral immune response mediated by circulating immunoglobulin
          defense response
          inflammatory response
          positive regulation of antibacterial peptide biosynthetic process
          positive regulation of biosynthetic process of antibacterial peptides active against Gram-positive bacteria
          signal transduction
          positive regulation of cell proliferation
          detection of biotic stimulus
          detection of bacterium
          maintenance of gastrointestinal epithelium
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          response to peptidoglycan
          response to muramyl dipeptide
          response to lipopolysaccharide
          detection of muramyl dipeptide
          negative regulation of interferon-gamma production
          negative regulation of interleukin-12 production
          negative regulation of interleukin-18 production
          negative regulation of interleukin-2 production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          negative regulation of toll-like receptor 2 signaling pathway
          activation of MAPK activity involved in innate immune response
          intracellular signal transduction
          defense response to bacterium
          regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          positive regulation of MAPK cascade
          positive regulation of phosphatidylinositol 3-kinase activity
          negative regulation of growth of symbiont in host
          innate immune response
          positive regulation of innate immune response
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of epithelial cell proliferation
          positive regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of phagocytosis
          defense response to Gram-positive bacterium
          positive regulation of B cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          positive regulation of oxidoreductase activity
          positive regulation of nitric-oxide synthase biosynthetic process
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          macrophage inflammatory protein-1 alpha production
          regulation of neutrophil chemotaxis
          positive regulation of cytokine production involved in inflammatory response
          negative regulation of macrophage apoptotic process
          positive regulation of prostaglandin-E synthase activity
          nucleotide binding
          protein binding
          ATP binding
          endodeoxyribonuclease activity, producing 5'-phosphomonoesters
          enzyme binding
          protein kinase binding
          muramyl dipeptide binding
          peptidoglycan binding
          CARD domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800007998.mm.1 NM_145857.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007998.mm.1 NM_145857.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007998.mm.1 TC0800001037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007999.mm.1 NM_145857.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007999.mm.1 NM_145857.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007999.mm.1 TC0800001037.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline