Search
Search
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035231.1 | - | 3 - 4 | 432 | 89 | |
| AK078773.1 | - | 4 - 5 | 622 | 89 | ||
| AK155752.1 | - | 3 - 4 | 444 | 89 | ||
| BC052163.1 | - | 4 - 5 | 583 | 6513848 | 89 | |
Set Membership: |
|