Search
Search

Tinagl1| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168333.1 | NP_001161805.1 | 9 - 10 | 1173 | 85 | |
| NM_023476.3 | NP_075965.2 | 9 - 10 | 1314 | 85 | ||
| GenBank mRNA | AB047402.1 | - | 9 - 10 | 1315 | 85 | |
| AB050626.1 | - | 9 - 10 | 1151 | 85 | ||
| AK087142.1 | - | 1174 | 85 | |||
| BC005738.1 | - | 9 - 10 | 1141 | 2650851 | 85 | |
| BC018539.1 | - | 9 - 10 | 1158 | 3596560 | 85 | |
| CT010275.1 | - | 8 - 9 | 932 | 85 | ||
Set Membership: |
|