Search
Search
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 10 - 11 | 1289 | 72 | |
| GenBank mRNA | AF237669.1 | - | 10 - 11 | 1248 | 72 | |
| AF239661.1 | - | 10 - 11 | 1290 | 72 | ||
| AK004876.1 | - | 9 - 10 | 1022 | 72 | ||
| AK053550.1 | - | 10 - 11 | 1276 | 72 | ||
| AK131140.1 | - | 795 | 72 | |||
| AK134654.1 | - | 1271 | 72 | |||
| AK165746.1 | - | 10 - 11 | 1283 | 72 | ||
Set Membership: |
|