Search
Search

Ero1lb| Gene Symbol: Ero1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026184.2 | NP_080460.2 | 4 - 5 | 546 | 74 | |
| GenBank mRNA | AF489856.1 | - | 4 - 5 | 352 | 74 | |
| AK004981.1 | - | 523 | 74 | |||
| AK006886.1 | - | 4 - 5 | 409 | 74 | ||
| AK041951.1 | - | 4 - 5 | 522 | 74 | ||
| AK160664.1 | - | 4 - 5 | 562 | 74 | ||
| BC058721.1 | - | 4 - 5 | 546 | 5705507 | 74 | |
| BC111900.1 | - | 4 - 5 | 516 | 40062137 | 74 | |
| BC112426.1 | - | 4 - 5 | 516 | 40062136 | 74 | |
| BC145888.1 | - | 4 - 5 | 497 | 40131191 | 74 | |
| BC145890.1 | - | 4 - 5 | 497 | 40131193 | 74 | |
Set Membership: |
|