Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470816_m1
          See other TMED9 GE Assays ›
          Gene Symbol:
          Tmed9
          Gene Name:
          transmembrane emp24 protein transport domain containing 9
          Gene Aliases:
          2400003B06Rik
          Chromosome Location:
          Chr.13: 55593135 - 55597697 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470816_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmed9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026211.3 NP_080487.2 3 - 4 451 68
          GenBank mRNA AK010259.1 - 3 - 4 425 68
          AK012474.1 - 3 - 4 444 68
          AK044120.1 - 3 - 4 312 68
          AK152496.1 - 3 - 4 444 68
          AK152581.1 - 3 - 4 444 68
          AK153383.1 - 3 - 4 444 68
          AK169001.1 - 3 - 4 424 68
          BC004691.1 - 3 - 4 435 3501295 68
          BC049282.1 - 3 - 4 417 2648375 68
          BC058801.1 - 3 - 4 417 6476565 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000667.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmed9

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          Golgi organization
          positive regulation of organelle organization
          protein transport
          COPI coating of Golgi vesicle
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055968 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 BC058801.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 CF382327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK153383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK169001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 BC058801.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 CF382327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK153383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK169001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129484 AK012474.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196635 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129484 AK012474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 BC058801.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 CF382327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK153383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK169001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK213802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 BC058801.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 CF382327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK153383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK169001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 BC058801.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 CF382327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK153383.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK169001.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 BC058801.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 CF382327.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK153383.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK169001.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29055968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142967 - -
          Whole Mouse Genome G4122A A_51_P262878 BC058801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 CF382327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK153383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK169001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK213802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 BC058801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 CF382327.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK153383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK169001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262878 AK213802.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129484 AK012474.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142967 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10534917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10534921 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29055967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29055968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29056039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10882960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10707784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10707776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10742349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10882962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10442647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10882966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10707783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10655675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29055976 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405489 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405492 - -
          Affymetrix HT MG-430 PM Array Plate 1448268_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439448_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405488 - -
          Mouse Genome 430 2.0 Array 1439448_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448268_at - -
          Mouse Genome 430 2.0 Array 1448268_PM_at - -
          Mouse Genome 430 2.0 Array 1439448_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67511_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67511_at - -
          Affymetrix Mouse Clariom S Array PSR1300004411.mm.1 NM_026211.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004411.mm.1 NM_026211.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004411.mm.1 TC1300000667.mm.1 MTA TC
          Mouse Expression Set 430 1448268_PM_at - -
          Mouse Expression Set 430 1448268_at - -
          Mouse Expression Set 430 1439448_PM_x_at - -
          Mouse Expression Set 430 1439448_x_at - -
          Murine Genome U74v2 Set 100074_at - -
          Murine Genome U74v2 Set 100074_PM_at - -
          Mouse Genome 430A 2.0 Array 1448268_PM_at - -
          Mouse Genome 430A 2.0 Array 1439448_x_at - -
          Mouse Genome 430A 2.0 Array 1448268_at - -
          Mouse Genome 430A 2.0 Array 1439448_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448268_at - -
          Affymetrix HT Mouse Genome 430A Array 1439448_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439448_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448268_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 740003 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2900068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720740 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720740 - -
          mouse-6 v1.1 (Illumina) scl44728.4.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 2900068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44728.4.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2981871 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720740 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2981871 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720740 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl44728.4.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline