Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471043_s1
          See other DDX43 GE Assays ›
          Gene Symbol:
          Ddx43 4930542C12Rik
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 43
          RIKEN cDNA 4930542C12 gene
          Gene Aliases:
          ENSMUSG00000070291, OTTMUSG00000019690
          -
          Chromosome Location:
          Chr.9: 78395777 - 78423589 on Build GRCm38
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00471043_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx43
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191044.1 NP_001177973.1 1 147
          Gene Symbol: 4930542C12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016017.1 - 149 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001052.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA secondary structure unwinding
          molecular_function
          ATP-dependent RNA helicase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26659368 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120542 - -
          Whole Mouse Genome G4122A A_52_P389017 AK006987.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389017 AK016017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389017 AK017030.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124900 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07642121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26659402 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1422215_PM_at - -
          Mouse Genome 430 2.0 Array 1422215_PM_at - -
          Mouse Genome 430 2.0 Array 1422215_at - -
          Affymetrix Mouse Clariom S Array PSR0900008287.mm.1 NM_001191044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008287.mm.1 NM_001191044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008287.mm.1 TC0900001052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008286.mm.1 NM_001191044.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008286.mm.1 NM_001191044.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008286.mm.1 TC0900001052.mm.1 MTA TC
          Mouse Expression Set 430 1422215_at - -
          Mouse Expression Set 430 1422215_PM_at - -
          Mouse Genome 430A 2.0 Array 1422215_at - -
          Mouse Genome 430A 2.0 Array 1422215_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422215_at - -
          Affymetrix HT Mouse Genome 430A Array 1422215_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 363974 - -
          ABI Mouse Genome Survey Microarray 420796 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2190093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605982 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605982 - -
          mouse-6 v1.1 (Illumina) scl35572.1.60_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35572.1.60_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190093 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline