Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471232_m1
          See other RIBC2 GE Assays ›
          Gene Symbol:
          Ribc2
          Gene Name:
          RIB43A domain with coiled-coils 2
          Gene Aliases:
          4930579A10Rik, Trib
          Chromosome Location:
          Chr.15: 85132060 - 85144569 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471232_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ribc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026357.2 NP_080633.2 2 - 3 364 117
          GenBank mRNA AK006801.1 - 2 - 3 361 117
          AK016311.1 - 2 - 3 365 117
          AK017028.1 - 2 - 3 384 117
          BC112373.1 - 2 - 3 289 40056860 117
          BY716712.1 - 2 - 3 384 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000803.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK016311.1,BC112373.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 AK016311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 AK017028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 BC112373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 NM_026357.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 AK016311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 AK017028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 BC112373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P264785 NM_026357.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138568 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155617 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154514 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 AK016311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 AK017028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 BC112373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 NM_026357.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 AK016311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 AK017028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 BC112373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P264785 NM_026357.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154514 - -
          Whole Mouse Genome G4122A A_51_P264785 AK016311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 AK017028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 BC112373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 NM_026357.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 AK016311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 AK017028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 BC112373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264785 NM_026357.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155617 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30398911 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425938 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425939 - -
          Affymetrix HT MG-430 PM Array Plate 1432086_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425936 - -
          Mouse Genome 430 2.0 Array 1432086_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432086_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67747_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67747_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500006323.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006323.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006323.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006330.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006330.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006330.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006322.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006322.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006322.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006324.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006324.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006324.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006329.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006329.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006329.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006328.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006328.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006328.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006325.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006325.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006325.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006327.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006327.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006327.mm.1 TC1500000803.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006321.mm.1 NM_026357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006321.mm.1 NM_026357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006321.mm.1 TC1500000803.mm.1 MTA TC
          Mouse Expression Set 430 1432086_a_at - -
          Mouse Expression Set 430 1432086_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432086_a_at - -
          Mouse Genome 430A 2.0 Array 1432086_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432086_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432086_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 303490 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253464 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline