Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471509_m1
          See other FTSJ2 GE Assays ›
          Gene Symbol:
          Ftsj2
          Gene Name:
          FtsJ homolog 2 (E. coli)
          Gene Aliases:
          2310037B18Rik
          Chromosome Location:
          Chr.5: 140327674 - 140331898 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471509_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ftsj2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026510.1 NP_080786.1 2 - 3 336 90
          GenBank mRNA AK009656.1 - 2 - 3 336 90
          AK017300.1 - 2 - 3 318 90
          AK145808.1 - 2 - 3 317 90
          BC064776.1 - 1 - 2 304 30094986 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003506.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ftsj2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          rRNA processing
          rRNA methylation
          methylation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140927 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK017300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK077929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK145808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 BC064776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 NM_026510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK017300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK077929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 AK145808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 BC064776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100242 NM_026510.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121952 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110549 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK017300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK077929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK145808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 BC064776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 NM_026510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK017300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK077929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 AK145808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 BC064776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100242 NM_026510.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24140924 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110549 - -
          Whole Mouse Genome G4122A A_52_P100242 AK017300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 AK077929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 AK145808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 BC064776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 NM_026510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 AK017300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 AK077929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 AK145808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 BC064776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100242 NM_026510.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114321 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04381758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04381762 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04701168 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04701160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04381760 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24140927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24140925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04332661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04381759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04701160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24140911 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535210 - -
          Affymetrix HT MG-430 PM Array Plate 1448969_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535208 - -
          Mouse Genome 430 2.0 Array 1448969_at - -
          Mouse Genome 430 2.0 Array 1448969_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68017_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68017_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500032150.mm.1 TC0500003506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032150.mm.1 NM_026510.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032150.mm.1 NM_026510.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032151.mm.1 TC0500003506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032151.mm.1 NM_026510.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032151.mm.1 NM_026510.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032153.mm.1 TC0500003506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032153.mm.1 NM_026510.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032153.mm.1 NM_026510.1 Refseq
          Mouse Expression Set 430 1448969_PM_at - -
          Mouse Expression Set 430 1448969_at - -
          Mouse Genome 430A 2.0 Array 1448969_at - -
          Mouse Genome 430A 2.0 Array 1448969_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448969_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448969_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 886765 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4540411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540411 - -
          Illumina Sentrix mouse-6.v1 bead array 4540411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25835.3_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2795174 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2795174 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline