Search
Search

Chid1| Gene Symbol: Chid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142681.1 | NP_001136153.1 | 4 - 5 | 521 | 71 | |
| NM_026522.5 | NP_080798.2 | 4 - 5 | 521 | 71 | ||
| GenBank mRNA | AK014071.1 | - | 4 - 5 | 470 | 71 | |
| AK014141.1 | - | 4 - 5 | 496 | 71 | ||
| AK036717.1 | - | 4 - 5 | 465 | 71 | ||
| AK050655.1 | - | 4 - 5 | 469 | 71 | ||
| AK147377.1 | - | 4 - 5 | 484 | 71 | ||
| AK160156.1 | - | 4 - 5 | 481 | 71 | ||
| AK160200.1 | - | 4 - 5 | 481 | 71 | ||
| BC006876.1 | - | 4 - 5 | 470 | 2647540 | 71 | |
| BC061063.1 | - | 4 - 5 | 481 | 6591338 | 71 | |
Set Membership: |
|