Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471893_g1
          See other 1600015I10RIK GE Assays ›
          Gene Symbol:
          1600015I10Rik
          Gene Name:
          RIKEN cDNA 1600015I10 gene
          Gene Aliases:
          AI429442, Doxl1
          Chromosome Location:
          Chr.6: 48929896 - 48933687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471893_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1600015I10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081273.2 NP_001074742.2 3 - 4 2185 120
          GenBank mRNA AK005467.1 - 2 - 3 299 120
          BC085280.1 - 3 - 4 2186 120
          BC099440.1 - 3 - 4 822 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000573.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 BC085280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 BC099440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 NM_001081273.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 AK005467.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 BC085280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 BC099440.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 NM_001081273.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P512061 AK005467.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970397 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151368 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 BC085280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 BC099440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 NM_001081273.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 AK005467.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 BC085280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 BC099440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 NM_001081273.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P512061 AK005467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 BC085280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 BC099440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 NM_001081273.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 AK005467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 BC085280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 BC099440.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 NM_001081273.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512061 AK005467.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05566848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05751176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05751178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05566845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05566842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05566846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05751186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24429996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24430083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05566849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05659836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05517443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05550902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24429999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05751184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05461068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05517451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538170 - -
          Affymetrix HT MG-430 PM Array Plate 1449327_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538169 - -
          Mouse Genome 430 2.0 Array 1449327_PM_at - -
          Mouse Genome 430 2.0 Array 1449327_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69761_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69761_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600004424.mm.1 TC0600000573.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004424.mm.1 NM_001081273.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004424.mm.1 NM_001081273.2 Refseq
          Mouse Expression Set 430 1449327_PM_at - -
          Mouse Expression Set 430 1449327_at - -
          Murine Genome U74v2 Set 131217_PM_at - -
          Murine Genome U74v2 Set 131217_at - -
          Mouse Genome 430A 2.0 Array 1449327_PM_at - -
          Mouse Genome 430A 2.0 Array 1449327_at - -
          Affymetrix HT Mouse Genome 430A Array 1449327_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449327_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316013 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222284 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222284 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline