Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471907_m1
          See other CTSLL3 GE Assays ›
          Gene Symbol:
          Ctsll3
          Gene Name:
          cathepsin L-like 3
          Gene Aliases:
          2310051M13Rik
          Chromosome Location:
          Chr.13: 60798250 - 60802844 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471907_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsll3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027344.3 NP_081620.2 6 - 7 877 73
          GenBank mRNA AK009935.1 - 1 - 2 272 73
          AK145382.1 - 6 - 7 881 73
          BC147423.1 - 5 - 6 816 9056481 73
          BC147424.1 - 5 - 6 816 9056499 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002139.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsll3

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          proteolysis involved in cellular protein catabolic process
          molecular_function
          cysteine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 AK145382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 NM_027344.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 AK009935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 AK145382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 NM_027344.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P238796 AK009935.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909337 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163279 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132540 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100696 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133084 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 AK145382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 NM_027344.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 AK009935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 AK145382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 NM_027344.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P238796 AK009935.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163279 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100696 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133084 - -
          Whole Mouse Genome G4122A A_51_P238796 AK145382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238796 NM_027344.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238796 AK009935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238796 AK145382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238796 NM_027344.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P238796 AK009935.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123562 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163279 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133084 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132540 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10792634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10792689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10792669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10792641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10792688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11082231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10605541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10540916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10792638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409855 - -
          Affymetrix HT MG-430 PM Array Plate 1421684_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409848 - -
          Mouse Genome 430 2.0 Array 1421684_at - -
          Mouse Genome 430 2.0 Array 1421684_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70202_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70202_at - -
          Affymetrix Mouse Clariom S Array PSR1300013719.mm.1 NM_027344.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013719.mm.1 NM_027344.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013719.mm.1 TC1300002139.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013716.mm.1 NM_027344.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013716.mm.1 NM_027344.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013716.mm.1 TC1300002139.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013718.mm.1 NM_027344.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013718.mm.1 NM_027344.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013718.mm.1 TC1300002139.mm.1 MTA TC
          Mouse Expression Set 430 1421684_PM_at - -
          Mouse Expression Set 430 1421684_at - -
          Mouse Genome 430A 2.0 Array 1421684_PM_at - -
          Mouse Genome 430A 2.0 Array 1421684_at - -
          Affymetrix HT Mouse Genome 430A Array 1421684_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421684_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 728015 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5690450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237270 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237270 - -
          mouse-6 v1.1 (Illumina) scl43867.3.1_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43867.3.1_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237270 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237270 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline