Search
Search

Fcrls| Gene Symbol: Fcrls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030707.3 | NP_109632.2 | 1 - 2 | 76 | 76 | |
| GenBank mRNA | AB028174.1 | - | 1 - 2 | 107 | 76 | |
| AF302046.1 | - | 1 - 2 | 145 | 76 | ||
| AF329486.1 | - | 1 - 2 | 77 | 76 | ||
| AK034125.1 | - | 1 - 2 | 77 | 76 | ||
| AK076123.1 | - | 1 - 2 | 110 | 76 | ||
| AY506556.1 | - | 1 - 2 | 116 | 76 | ||
| AY506557.1 | - | 1 - 2 | 116 | 76 | ||
| BC016551.1 | - | 1 - 2 | 119 | 3602803 | 76 | |
Set Membership: |
|