Search
Search
| Gene Symbol: Pum2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160219.1 | NP_001153691.1 | 8 - 9 | 1246 | 76 | |
| NM_001160220.1 | NP_001153692.1 | 8 - 9 | 1246 | 76 | ||
| NM_001160221.1 | NP_001153693.1 | 8 - 9 | 1061 | 76 | ||
| NM_001160222.1 | NP_001153694.1 | 8 - 9 | 1061 | 76 | ||
| NM_030723.2 | NP_109648.2 | 8 - 9 | 1170 | 76 | ||
| NR_027670.1 | 10 - 11 | 1445 | 76 | |||
| GenBank mRNA | AF315590.1 | - | 8 - 9 | 1186 | 76 | |
| AK048119.1 | - | 3 - 4 | 352 | 76 | ||
| AK122225.1 | - | 10 - 11 | 1331 | 76 | ||
| AK134185.1 | - | 8 - 9 | 1061 | 76 | ||
| AK141682.1 | - | 8 - 9 | 1108 | 76 | ||
| AK147617.1 | - | 10 - 11 | 1430 | 76 | ||
| AK163964.1 | - | 8 - 9 | 1036 | 76 | ||
| AY027917.1 | - | 8 - 9 | 1086 | 76 | ||
| BC041773.1 | - | 7 - 8 | 1035 | 3986207 | 76 | |
Set Membership: |
|