Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473479_m1
          See other ENPP5 GE Assays ›
          Gene Symbol:
          Enpp5
          Gene Name:
          ectonucleotide pyrophosphatase/phosphodiesterase 5
          Gene Aliases:
          AW061004, D17Abb1e, E-NPP 5, mKIAA4152
          Chromosome Location:
          Chr.17: 44078796 - 44086567 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473479_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enpp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032003.2 NP_114392.1 1 - 2 140 66
          GenBank mRNA AF233377.1 - 1 - 2 117 66
          AK220510.1 - 1 - 2 83 66
          BC138977.1 - 1 - 2 55 8861999 66
          BC138978.1 - 1 - 2 55 8862000 66
          BC145484.1 - 1 - 2 55 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000842.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF233377.1,BC138977.1,BC138978.1,NM_032003.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Enpp5

          Gene Ontology Categories:

          Function(s) Process(es)

          cell communication
          metabolic process
          nucleotide catabolic process
          catalytic activity
          nuclease activity
          nucleotide diphosphatase activity
          hydrolase activity
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105074 AK032341.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105074 AK220510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105074 BC089320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105074 NM_032003.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105074 AF233377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021704 BC089320.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25734 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166309 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105074 AK032341.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105074 AK220510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105074 BC089320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105074 NM_032003.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105074 AF233377.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362104 AF233377.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31192840 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03320445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166309 - -
          Whole Mouse Genome G4122A A_51_P362104 AF233377.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105074 AK032341.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105074 AK220510.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105074 BC089320.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105074 NM_032003.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105074 AF233377.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13274461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13297437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13297442 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13274465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13274457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31192847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13143686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13138495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13159961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13297302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13159965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13143681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13297437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13297318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13262847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13138496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13297311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13112023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13590897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13274456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13123144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31192878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445345 - -
          Affymetrix HT MG-430 PM Array Plate 1425702_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445338 - -
          Mouse Genome 430 2.0 Array 1425702_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425702_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83965_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83965_at - -
          Affymetrix Mouse Clariom S Array PSR1700008090.mm.1 NM_001168620.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008090.mm.1 NM_032003.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008090.mm.1 TC1700000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008087.mm.1 NM_032003.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008087.mm.1 TC1700000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008088.mm.1 NM_032003.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008088.mm.1 TC1700000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008091.mm.1 NM_001168620.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008091.mm.1 NM_032003.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008091.mm.1 TC1700000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008089.mm.1 NM_001168620.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008089.mm.1 NM_032003.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008089.mm.1 TC1700000842.mm.1 MTA TC
          Mouse Expression Set 430 1425702_PM_a_at - -
          Mouse Expression Set 430 1425702_a_at - -
          Mouse Genome 430A 2.0 Array 1425702_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425702_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425702_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425702_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 370857 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217118 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217118 - -
          mouse-6 v1.1 (Illumina) scl50698.5_378-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50698.5_378-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217118 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217118 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50698.5_378-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline