Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473571_g1
          See other SVAL2 GE Assays ›
          Gene Symbol:
          Sval2
          Gene Name:
          seminal vesicle antigen-like 2
          Gene Aliases:
          SLP-M
          Chromosome Location:
          Chr.6: 41860339 - 41864322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473571_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sval2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032542.1 NP_115931.1 3 - 4 309 107
          GenBank mRNA AB050614.1 - 3 - 4 309 107
          BC119343.1 - 3 - 4 409 8734092 107
          BC119345.1 - 3 - 4 409 8734094 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000447.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sval2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954577 NM_032542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954577 AB050614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954577 BC119343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954577 BC119345.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556094 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127906 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954577 NM_032542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954577 AB050614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954577 BC119343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954577 BC119345.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24396110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127906 - -
          Whole Mouse Genome G4122A A_51_P128010 NM_032542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128010 AB050614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128010 BC119343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P128010 BC119345.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135208 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05675037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24396020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05675028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05711753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05711751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05047690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05675018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05675016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05294108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05207634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05711740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05711748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05047663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24396124 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537688 - -
          Affymetrix HT MG-430 PM Array Plate 1450809_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10537684 - -
          Mouse Genome 430 2.0 Array 1450809_PM_at - -
          Mouse Genome 430 2.0 Array 1450809_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84543_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84543_at - -
          Affymetrix Mouse Clariom S Array PSR0600003572.mm.1 NM_032542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003572.mm.1 NM_032542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003572.mm.1 TC0600000447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003574.mm.1 TC0600000447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003573.mm.1 NM_032542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003573.mm.1 NM_032542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003573.mm.1 TC0600000447.mm.1 MTA TC
          Mouse Expression Set 430 1450809_PM_at - -
          Mouse Expression Set 430 1450809_at - -
          Mouse Genome 430A 2.0 Array 1450809_PM_at - -
          Mouse Genome 430A 2.0 Array 1450809_at - -
          Affymetrix HT Mouse Genome 430A Array 1450809_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450809_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 638314 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160610 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160610 - -
          mouse-6 v1.1 (Illumina) GI_14211543-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160610 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160610 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline