Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473644_m1
          See other ARPP21 GE Assays ›
          Gene Symbol:
          Arpp21
          Gene Name:
          cyclic AMP-regulated phosphoprotein, 21
          Gene Aliases:
          0710001E13Rik, AI853636, ARPP-21, D9Bwg1012e, R3hdm3, Tarpp
          Chromosome Location:
          Chr.9: 112065091 - 112236022 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473644_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arpp21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177616.1 NP_001171087.1 16 - 17 2213 75
          NM_001177617.1 NP_001171088.1 13 - 14 2128 75
          NM_001177618.1 NP_001171089.1 16 - 17 1971 75
          NM_033264.2 NP_150289.1 16 - 17 2100 75
          GenBank mRNA AF324451.1 - 16 - 17 2060 75
          AK032417.1 - 16 - 17 75
          AK032543.1 - 14 - 15 1781 75
          AK034376.1 - 15 - 16 1819 75
          AK137974.1 - 13 - 14 2129 75
          BC053001.1 - 15 - 16 1937 5686446 75
          BC098368.1 - 16 - 17 4437 75
          BC128376.1 - 15 - 16 1819 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003138.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arpp21

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to heat
          molecular_function
          nucleic acid binding
          calmodulin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177618.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_001177618.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_033264.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 NM_033264.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026555.mm.1 TC0900003138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026554.mm.1 NM_001177617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026554.mm.1 NM_001177617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026554.mm.1 TC0900003138.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline