Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474538_s1
          See other PCDHB18 GE Assays ›
          Gene Symbol:
          Pcdhb18
          Gene Name:
          protocadherin beta 18
          Gene Aliases:
          Pcdhb9, PcdhbR
          Chromosome Location:
          Chr.18: 37489465 - 37494503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474538_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053143.2 NP_444373.1 1 509 104
          GenBank mRNA AK140341.1 - 1 510 104
          AY013779.1 - 1 355 104
          BC099958.1 - 1 445 30086462 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000375.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb18

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184796 NM_053143.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184796 AK140341.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176333 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131094 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184796 NM_053143.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184796 AK140341.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511918 AY013779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511918 BC099958.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511918 NM_053143.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P511918 AK140341.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628600 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131094 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08968311 - -
          Whole Mouse Genome G4122A A_51_P511918 AY013779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511918 BC099958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511918 NM_053143.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P511918 AK140341.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121090 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009091 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455119 - -
          Affymetrix HT MG-430 PM Array Plate 1421723_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455118 - -
          Mouse Genome 430 2.0 Array 1421723_at - -
          Mouse Genome 430 2.0 Array 1421723_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93889_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93889_at - -
          Affymetrix Mouse Clariom S Array PSR1800002861.mm.1 NM_053143.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002861.mm.1 NM_053143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002861.mm.1 TC1800000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002864.mm.1 NM_053143.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002864.mm.1 NM_053143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002864.mm.1 TC1800000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002862.mm.1 NM_053143.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002862.mm.1 NM_053143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002862.mm.1 TC1800000375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002863.mm.1 NM_053143.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002863.mm.1 NM_053143.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002863.mm.1 TC1800000375.mm.1 MTA TC
          Mouse Expression Set 430 1421723_at - -
          Mouse Expression Set 430 1421723_PM_at - -
          Mouse Genome 430A 2.0 Array 1421723_PM_at - -
          Mouse Genome 430A 2.0 Array 1421723_at - -
          Affymetrix HT Mouse Genome 430A Array 1421723_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421723_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381426 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643460 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643460 - -
          mouse-6 v1.1 (Illumina) scl52053.1.1_294-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52053.1.1_294-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643460 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643460 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline