Search
Search

Cadm3| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053199.3 | NP_444429.1 | 4 - 5 | 718 | 62 | |
| GenBank mRNA | AF195662.1 | - | 4 - 5 | 662 | 62 | |
| AK036352.1 | - | 5 - 6 | 774 | 62 | ||
| AK038917.1 | - | 4 - 5 | 675 | 62 | ||
| AK053077.1 | - | 4 - 5 | 672 | 62 | ||
| AY059393.1 | - | 4 - 5 | 517 | 62 | ||
| BC029659.1 | - | 1 - 2 | 137 | 62 | ||
| BC115417.2 | - | 4 - 5 | 623 | 40093410 | 62 | |
| BC115418.2 | - | 4 - 5 | 623 | 40093417 | 62 | |
Set Membership: |
|