Search
Search
| Gene Symbol: Tle6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053254.2 | NP_444484.1 | 11 - 12 | 1550 | 77 | |
| GenBank mRNA | AB221587.1 | - | 10 - 11 | 1411 | 77 | |
| AF145957.2 | - | 11 - 12 | 1550 | 77 | ||
| AK007880.1 | - | 5 - 6 | 782 | 77 | ||
| AK154391.1 | - | 8 - 9 | 1511 | 77 | ||
| BC023315.1 | - | 11 - 12 | 1449 | 77 | ||
| BC048832.1 | - | 8 - 9 | 1389 | 4506029 | 77 | |
Set Membership: |
|