Search
Search

Impa2| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053261.2 | NP_444491.1 | 1 - 2 | 288 | 99 | |
| GenBank mRNA | AF353730.1 | - | 1 - 2 | 287 | 99 | |
| AK154489.1 | - | 1 - 2 | 326 | 99 | ||
| AK154845.1 | - | 1 - 2 | 259 | 99 | ||
| AK170962.1 | - | 1 - 2 | 326 | 99 | ||
| AK171626.1 | - | 1 - 2 | 261 | 99 | ||
| BC011093.1 | - | 1 - 2 | 263 | 4012163 | 99 | |
Set Membership: |
|