Search
Search

Apoa5| Gene Symbol: Apoa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080434.3 | NP_536682.2 | 1 - 2 | 64 | 66 | |
| GenBank mRNA | AF327059.1 | - | 1 - 2 | 64 | 66 | |
| AK004903.1 | - | 1 - 2 | 67 | 66 | ||
| AK004936.1 | - | 1 - 2 | 64 | 66 | ||
| AK050280.1 | - | 1 - 2 | 65 | 66 | ||
| BC011198.1 | - | 2 - 3 | 90 | 4196296 | 66 | |
| BC029334.1 | - | 1 - 2 | 79 | 66 | ||
Set Membership: |
|