Search
Search

Smc1b| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080470.1 | NP_536718.1 | 12 - 13 | 2077 | 143 | |
| GenBank mRNA | AF303827.1 | - | 12 - 13 | 2077 | 143 | |
| BC129800.1 | - | 12 - 13 | 2068 | 40048794 | 143 | |
Set Membership: |
|