Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475829_g1
          See other AGRP GE Assays ›
          Gene Symbol:
          Agrp
          Gene Name:
          agouti related neuropeptide
          Gene Aliases:
          Agrt, Art
          Chromosome Location:
          Chr.8: 105566695 - 105579845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475829_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Agrp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271806.1 NP_001258735.1 3 - 4 447 86
          NM_007427.3 NP_031453.1 3 - 4 337 86
          GenBank mRNA BC079902.1 - 3 - 4 334 30611244 86
          CO044337.1 - 2 - 3 86
          U89484.1 - 1 - 2 127 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003246.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Agrp

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neuropeptide signaling pathway
          adult feeding behavior
          hormone-mediated signaling pathway
          eating behavior
          regulation of feeding behavior
          positive regulation of feeding behavior
          receptor binding
          neuropeptide hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26134617 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26134616 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049717 BC079902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049717 AK138796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049717 NM_007427.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157442 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141623 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136873 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165993 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170747 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049717 BC079902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049717 AK138796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049717 NM_007427.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347965 BC079902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347965 U89484.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347965 AK138796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347965 NM_007427.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26134626 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141623 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06627031 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01818076 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06627003 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01818117 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06627016 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170747 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136873 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165993 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01818058 - -
          Whole Mouse Genome G4122A A_51_P347965 BC079902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347965 U89484.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347965 AK138796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347965 NM_007427.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125036 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112605 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01818058 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170747 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165993 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06627016 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141623 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136873 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07146712 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06928387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07146713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06928375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07146715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07146714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06928393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06928381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06928404 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26134616 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26134617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26134595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07288710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06933374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06805666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06928392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07154451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07348798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06939295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06928398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07154450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06861690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07348796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07146712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26134657 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581301 - -
          Affymetrix HT MG-430 PM Array Plate 1421690_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581299 - -
          Mouse Genome 430 2.0 Array 1421690_s_at - -
          Mouse Genome 430 2.0 Array 1421690_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11604_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800021759.mm.1 NM_001271806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021759.mm.1 NM_007427.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021759.mm.1 TC0800003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021762.mm.1 NM_007427.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021762.mm.1 TC0800003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021764.mm.1 NM_001271806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021764.mm.1 TC0800003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021760.mm.1 NM_001271806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021760.mm.1 NM_007427.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021760.mm.1 TC0800003246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021761.mm.1 NM_001271806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021761.mm.1 NM_007427.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021761.mm.1 TC0800003246.mm.1 MTA TC
          Mouse Expression Set 430 1421690_PM_s_at - -
          Mouse Expression Set 430 1421690_s_at - -
          Murine Genome U74v2 Set 99399_PM_at - -
          Murine Genome U74v2 Set 99399_at - -
          Mouse Genome 430A 2.0 Array 1421690_s_at - -
          Mouse Genome 430A 2.0 Array 1421690_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421690_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421690_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 796850 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1990082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703427 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703427 - -
          mouse-6 v1.1 (Illumina) scl34398.4.1_108-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34398.4.1_108-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2703427 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2703427 - -

          Back To Top
           

          Citations (1)

          Fibroblast growth factor 21 corrects obesity in mice.
          Authors
          Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
          Journal
          Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 UTC 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline