Search
Search

Barx1| Gene Symbol: Barx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007526.4 | NP_031552.2 | 2 - 3 | 595 | 74 | |
| GenBank mRNA | AB221538.1 | - | 1 - 2 | 193 | 74 | |
| AJ297677.1 | - | 2 - 3 | 622 | 74 | ||
| BC119047.1 | - | 1 - 2 | 274 | 74 | ||
| BC119049.1 | - | 1 - 2 | 274 | 74 | ||
| Y07960.1 | - | 2 - 3 | 337 | 74 | ||
Set Membership: |
|